Merck
Elevated serum ferritin.
JAMA (2014-08-21)
Lisa B VanWagner, Richard M Green
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL