Merck
  • Histone methyltransferase EZH2 is transcriptionally induced by estradiol as well as estrogenic endocrine disruptors bisphenol-A and diethylstilbestrol.
Histone methyltransferase EZH2 is transcriptionally induced by estradiol as well as estrogenic endocrine disruptors bisphenol-A and diethylstilbestrol.
Journal of molecular biology (2014-08-05)
Arunoday Bhan, Imran Hussain, Khairul I Ansari, Samara A M Bobzean, Linda I Perrotti, Subhrangsu S Mandal
ABSTRAKT
Enhancer of Zeste homolog 2 (EZH2), a methyltransferase specific to histone 3 lysine 27, is a critical player in gene silencing and is overexpressed in breast cancer. Our studies demonstrate that EZH2 is transcriptionally induced by estradiol in cultured breast cancer cells and in the mammary glands of ovariectomized rats. EZH2 promoter contains multiple functional estrogen-response elements. Estrogen receptors (ERs) and ER coregulators such as mixed lineage leukemia (MLL) histone methylases (MLL2 and MLL3) and histone acetyltransferase CBP/P300 bind to the EZH2 promoter in the presence of estradiol and regulate estradiol-induced EZH2 expression. EZH2 expression is also increased upon exposure to estrogenic endocrine disrupting chemicals (EDCs) such as bisphenol-A (BPA) and diethylstilbestrol (DES). Similar to estradiol, BPA and DES-induced EZH2 expression is coordinated by ERs, MLLs and CBP/P300. In summary, we demonstrate that EZH2 is transcriptionally regulated by estradiol in vitro and in vivo, and its expression is potentially dysregulated upon exposure to estrogenic EDCs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethylstilbestrol, ≥99% (HPLC)
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, analytical standard
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications