Merck
Two types of overcontraction are involved in intrarenal artery dysfunction in type II diabetic mouse.
The Journal of pharmacology and experimental therapeutics (2014-08-03)
Koji Nobe, Yasuhiro Takenouchi, Keizo Kasono, Terumasa Hashimoto, Kazuo Honda
ABSTRAKT
Contractile responses in small intrarenal arteries are associated with diabetic nephropathy. However, the mechanisms that induce and maintain altered small vessel contraction are not clearly understood. To further understand intrarenal artery dysfunction in diabetes, phenylephrine (PE)-induced force development was assessed in the intrarenal artery [interlobar artery (ILA)] in control (lean) and type II diabetic (ob/ob) mice. PE-induced dose-dependent force development in the ILA was significantly greater in ob/ob mice than in lean mice (592.8 ± 5.2 and 770.1 ± 12.1 µ/mm tissue, respectively, following administration of 30 µM PE, n = 5). Under high-glucose conditions (twice the normal concentration of glucose), PE-induced force development in the ILA was only enhanced in ob/ob mice (946.0 ± 18.2 µN/mm tissue; n = 5). ILA dysfunction reduces blood flow to the glomerulus and may induce diabetic nephropathy. Basal overcontraction of the ILA in ob/ob mice under normal-glucose conditions was reduced by pretreatment with rottlerin, a calcium-independent protein kinase C (PKCδ) inhibitor. Total PKC activity was also reduced by rottlerin. Under high-glucose conditions, the enhanced ILA contraction in diabetic mice was suppressed by rho A and rho kinase inhibitors. Our results indicate two types of ILA dysfunction in diabetes, as follows: 1) a basal increase in PE-induced contraction under normal-glucose conditions, and 2) extracellular glucose-dependent enhancement of PE-induced contraction. We believe that these dysfunctions are mediated by the activation of the PKCδ and rho A-rho kinase pathways, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard