Merck
  • The P600-as-P3 hypothesis revisited: single-trial analyses reveal that the late EEG positivity following linguistically deviant material is reaction time aligned.
The P600-as-P3 hypothesis revisited: single-trial analyses reveal that the late EEG positivity following linguistically deviant material is reaction time aligned.
Brain and language (2014-08-26)
Jona Sassenhagen, Matthias Schlesewsky, Ina Bornkessel-Schlesewsky
ABSTRAKT
The P600, a late positive ERP component following linguistically deviant stimuli, is commonly seen as indexing structural, high-level processes, e.g. of linguistic (re)analysis. It has also been identified with the P3 (P600-as-P3 hypothesis), which is thought to reflect a systemic neuromodulator release facilitating behavioural shifts and is usually response time aligned. We investigated single-trial alignment of the P600 to response, a critical prediction of the P600-as-P3 hypothesis. Participants heard sentences containing morphosyntactic and semantic violations and responded via a button press. The elicited P600 was perfectly response aligned, while an N400 following semantic deviations was stimulus aligned. This is, to our knowledge, the first single-trial analysis of language processing data using within-sentence behavioural responses as temporal covariates. Results support the P600-as-P3 perspective and thus constitute a step towards a neurophysiological grounding of language-related ERPs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Norepinephrine hydrochloride, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
DL-Norepinephrine hydrochloride, crystalline, ≥97% (TLC)
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt