Merck
Levobupivacaine inhibits lipopolysaccharide-induced high mobility group box 1 release in vitro and in vivo.
The Journal of surgical research (2014-07-16)
Yunfen Ge, Shuangfei Hu, Yunlong Zhang, Wenyuan Wang, Qiong Xu, Leping Zhou, Hui Mao
ABSTRAKT
The aim of the study was to investigate whether levobupivacaine (LB) suppressed lipopolysaccharide (LPS)-induced high mobility group box 1 (HMGB1) release in vitro and in vivo, and to determin its molecular mechanisms of action. RAW264.7 cells were treated with LPS and LB for 24 h. Levels of HMGB1, nuclear factor-kappa B (NF-κB) and phosphorylated p38 mitogen-activated protein kinase (MAPK) were measured by Enzyme-linked immunosorbent assay and Western blotting; the levels of HMGB1 messenger RNA were measured by real-time polymerase chain reaction. In addition, cecal ligation and puncture-induced septic C57BL/6 received LB infusion, and the levels of HMGB1 and functional parameters of multiple organs determined using several detection kits. LB inhibited HMGB1 release in vitro and in vivo. Furthermore, LB inhibited the translocation of NF-κB and phosphorylation of p38 MAPK in vitro. Mice treated with LB infusion improved survival in mice and significantly reduced cecal ligation and puncture-induced dysfunction of organs. LB suppresses LPS-induced HMGB1 release in vitro and in vivo by partially inhibiting NF-κB/p38 MAPK pathways. LB can rescue mice from sepsis and protect against organ dysfunction in septic mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Bupivacaine hydrochloride, European Pharmacopoeia (EP) Reference Standard