Merck
Thymidine phosphorylase participates in platelet signaling and promotes thrombosis.
Circulation research (2014-10-08)
Wei Li, Alba Gigante, Maria-Jesus Perez-Perez, Hong Yue, Michio Hirano, Thomas M McIntyre, Roy L Silverstein
ABSTRAKT
Platelets contain abundant thymidine phosphorylase (TYMP), which is highly expressed in diseases with high risk of thrombosis, such as atherosclerosis and type II diabetes mellitus. To test the hypothesis that TYMP participates in platelet signaling and promotes thrombosis. By using a ferric chloride (FeCl3)-induced carotid artery injury thrombosis model, we found time to blood flow cessation was significantly prolonged in Tymp(-/-) and Tymp(+/-) mice compared with wild-type mice. Bone marrow transplantation and platelet transfusion studies demonstrated that platelet TYMP was responsible for the antithrombotic phenomenon in the TYMP-deficient mice. Collagen-, collagen-related peptide-, adenosine diphosphate-, or thrombin-induced platelet aggregation were significantly attenuated in Tymp(+/-) and Tymp(-/-) platelets, and in wild type or human platelets pretreated with TYMP inhibitor KIN59. Tymp deficiency also significantly decreased agonist-induced P-selectin expression. TYMP contains an N-terminal SH3 domain-binding proline-rich motif and forms a complex with the tyrosine kinases Lyn, Fyn, and Yes in platelets. TYMP-associated Lyn was inactive in resting platelets, and TYMP trapped and diminished active Lyn after collagen stimulation. Tymp/Lyn double haploinsufficiency diminished the antithrombotic phenotype of Tymp(+/-) mice. TYMP deletion or inhibition of TYMP with KIN59 dramatically increased platelet-endothelial cell adhesion molecule 1 tyrosine phosphorylation and diminished collagen-related peptide- or collagen-induced AKT phosphorylation. In vivo administration of KIN59 significantly inhibited FeCl3-induced carotid artery thrombosis without affecting hemostasis. TYMP participates in multiple platelet signaling pathways and regulates platelet activation and thrombosis. Targeting TYMP might be a novel antiplatelet and antithrombosis therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran