Merck
Valproic Acid treatment after experimental subarachnoid hemorrhage.
Acta neurochirurgica. Supplement (2014-11-05)
Michael K Tso, Elliot Lass, Jinglu Ai, R Loch Macdonald
ABSTRAKT
Subarachnoid hemorrhage (SAH) can result in significant brain injury. Valproic acid (VPA), a widely-used anti-epileptic drug, was investigated as a possible neuroprotective drug in a prechiasmatic injection model of SAH in mice. Mice were randomized to the following experimental groups: SAH, SAH + VPA, Sham, and Sham + VPA. VPA (400 mg/kg) or saline was administered within 30 min of SAH induction and every 12 h thereafter for 48 h. Neurobehavioral assessments using the modified Garcia Score were conducted at 24 and 48 h. Brain injury was assessed at 48 h with fluoro-jade b and caspase-3/NeuN histo- and immunohistochemistry. Vasospasm was assessed in the MCA branches using hematoxylin & eosin histology. SAH mice treated with VPA appeared to have improved neurobehavioral assessments at both 24 and 48 h compared to untreated SAH mice. VPA treatment in SAH mice also significantly decreased the number of degenerating neurons on fluoro-jade b staining. In VPA-treated SAH mice, there was a trend toward a decrease in the number of apoptotic neurons on caspase-3/NeuN immunohistochemistry. VPA did not significantly affect vasospasm. This study demonstrated that VPA improves neurological outcome and decreases brain injury in a mouse model of SAH.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Propylpentanoic acid
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Kwas walproinowy, United States Pharmacopeia (USP) Reference Standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®