Merck
  • Prophylactic effects of elastin peptide derived from the bulbus arteriosus of fish on vascular dysfunction in spontaneously hypertensive rats.
Prophylactic effects of elastin peptide derived from the bulbus arteriosus of fish on vascular dysfunction in spontaneously hypertensive rats.
Life sciences (2014-12-03)
Kumiko Takemori, Ei Yamamoto, Hiroyuki Ito, Takashi Kometani
ABSTRAKT
To determine the prophylactic effects of an elastin peptide derived from the bulbus arteriosus of bonitos and prolylglycine (PG), a degradation product of elastin peptide, on vascular dysfunction in spontaneously hypertensive rats (SHRs). Male 15-week-old SHR/Izm rats were fed without (control group) or with elastin peptide (1 g/kg body weight) for 5 weeks (EP group), or were infused via an osmotic mini-pump for 4 weeks with PG (PG group) or saline (control group). Using thoracic aortas, we assessed endothelial changes by scanning electron microscopy. Vascular reactivity (contraction and relaxation) and pressure-induced distension was compared. mRNA production levels of endothelial nitric oxide synthase (eNOS) and intercellular adhesion molecule-1 (ICAM-1) were investigated by real-time-polymerase chain reaction. Aortas of the EP group displayed limited endothelial damage compared with that in the control group. Under treatment of SHRs with elastin peptide, the effect of phenylephrine returned closer to the normal level observed in normotensive Wistar-Kyoto (WKY/Izm) rats. mRNA production of eNOS (but not ICAM-1) was greater in the EP group than in the control group. Endothelial damage was suppressed and pressure-induced vascular distension was greater in the PG group than in the corresponding control group. These results suggest that elastin peptide from bonitos elicits prophylactic affects hypertension-associated vascular dysfunction by targeting the eNOS signaling pathway. PG may be a key mediator of the beneficial effects of elastin peptide.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Elastin, soluble from bovine neck ligament, salt-free, lyophilized powder
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Pro-Gly
Sigma-Aldrich
1,3-Propanediol, 98%