Merck
The neuroprotective actions of oestradiol and oestrogen receptors.
Nature reviews. Neuroscience (2014-11-27)
Maria-Angeles Arevalo, Iñigo Azcoitia, Luis M Garcia-Segura
ABSTRAKT
Hormones regulate homeostasis by communicating through the bloodstream to the body's organs, including the brain. As homeostatic regulators of brain function, some hormones exert neuroprotective actions. This is the case for the ovarian hormone 17β-oestradiol, which signals through oestrogen receptors (ERs) that are widely distributed in the male and female brain. Recent discoveries have shown that oestradiol is not only a reproductive hormone but also a brain-derived neuroprotective factor in males and females and that ERs coordinate multiple signalling mechanisms that protect the brain from neurodegenerative diseases, affective disorders and cognitive decline.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, analytical standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications