Merck
Dose-dependent nuclear β-catenin response segregates endomesoderm along the sea star primary axis.
Development (Cambridge, England) (2014-12-18)
Brenna S McCauley, Eda Akyar, H Rosa Saad, Veronica F Hinman
ABSTRAKT
In many invertebrates, the nuclearization of β-catenin at one pole of the embryo initiates endomesoderm specification. An intriguing possibility is that a gradient of nuclear β-catenin (nβ-catenin), similar to that operating in vertebrate neural tube patterning, functions to distinguish cell fates in invertebrates. To test this hypothesis, we determined the function of nβ-catenin during the early development of the sea star, which undergoes a basal deuterostomal mode of embryogenesis. We show that low levels of nβ-catenin activity initiate bra, which is expressed in the future posterior endoderm-fated territory; intermediate levels are required for expression of foxa and gata4/5/6, which are later restricted to the endoderm; and activation of ets1 and erg in the mesoderm-fated territory requires the highest nβ-catenin activity. Transcription factors acting downstream of high nβ-catenin segregate the endoderm/mesoderm boundary, which is further reinforced by Delta/Notch signaling. Significantly, therefore, in sea stars, endomesoderm segregation arises through transcriptional responses to levels of nβ-catenin activity. Here, we describe the first empirical evidence of a dose-dependent response to a dynamic spatiotemporal nβ-catenin activity that patterns cell fates along the primary axis in an invertebrate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Lithium chloride solution, 2 M in ethanol
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Supelco
Roztwór elektrolitu, nonaqueous, LiCl in ethanol (saturated)