Merck
Bortezomib-based therapy for newly diagnosed mantle-cell lymphoma.
The New England journal of medicine (2015-03-05)
Tadeusz Robak, Huiqiang Huang, Jie Jin, Jun Zhu, Ting Liu, Olga Samoilova, Halyna Pylypenko, Gregor Verhoef, Noppadol Siritanaratkul, Evgenii Osmanov, Julia Alexeeva, Juliana Pereira, Johannes Drach, Jiri Mayer, Xiaonan Hong, Rumiko Okamoto, Lixia Pei, Brendan Rooney, Helgi van de Velde, Franco Cavalli
ABSTRAKT
The proteasome inhibitor bortezomib was initially approved for the treatment of relapsed mantle-cell lymphoma. We investigated whether substituting bortezomib for vincristine in frontline therapy with R-CHOP (rituximab, cyclophosphamide, doxorubicin, vincristine, and prednisone) could improve outcomes in patients with newly diagnosed mantle-cell lymphoma. In this phase 3 trial, we randomly assigned 487 adults with newly diagnosed mantle-cell lymphoma who were ineligible or not considered for stem-cell transplantation to receive six to eight 21-day cycles of R-CHOP intravenously on day 1 (with prednisone administered orally on days 1 to 5) or VR-CAP (R-CHOP regimen, but replacing vincristine with bortezomib at a dose of 1.3 mg per square meter of body-surface area on days 1, 4, 8, and 11). The primary end point was progression-free survival. After a median follow-up of 40 months, median progression-free survival (according to independent radiologic review) was 14.4 months in the R-CHOP group versus 24.7 months in the VR-CAP group (hazard ratio favoring the VR-CAP group, 0.63; P<0.001), a relative improvement of 59%. On the basis of investigator assessment, the median durations of progression-free survival were 16.1 months and 30.7 months, respectively (hazard ratio, 0.51; P<0.001), a relative improvement of 96%. Secondary end points were consistently improved in the VR-CAP group, including the complete response rate (42% vs. 53%), the median duration of complete response (18.0 months vs. 42.1 months), the median treatment-free interval (20.5 months vs. 40.6 months), and the 4-year overall survival rate (54% vs. 64%). Rates of neutropenia and thrombocytopenia were higher in the VR-CAP group. VR-CAP was more effective than R-CHOP in patients with newly diagnosed mantle-cell lymphoma but at the cost of increased hematologic toxicity. (Funded by Janssen Research and Development and Millennium Pharmaceuticals; LYM-3002 ClinicalTrials.gov number, NCT00722137.).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals