Merck
Clinical and molecular implications of MED15 in head and neck squamous cell carcinoma.
The American journal of pathology (2015-03-21)
Zaki Shaikhibrahim, Anne Offermann, Rebecca Halbach, Wenzel Vogel, Martin Braun, Glen Kristiansen, Friedrich Bootz, Jörg Wenzel, Ralf Mikut, Claudia Lengerke, Markus Reischl, Andreas Schröck, Sven Perner
ABSTRAKT
Head and neck squamous cell carcinoma (HNSCC) progression depends on various dysregulated pathways. Regulation of diverse pathways is mediated by the mediator complex. The mediator subunit MED15 is essential for transforming growth factor (TGF)-β signaling and involved in breast and prostate cancers. We investigated the implication of MED15 in HNSCC. IHC for MED15 was performed on 324 tissue samples, and TGF-β assessed the use of Ki-67 and pSMAD3 as markers. MED15 knockdown followed by proliferation and migration assays, as well as TGF-β1 treatment followed by MED15 analysis, was also performed. MED15 was overexpressed in 35% of primary tumors, 30% of lymph node metastases, and 70% of recurrences in contrast to no or low expression in benign tumors. MED15 overexpression in primary tumors from patients who developed recurrences was associated with higher mortality rates and occurred at highest frequency in oral cavity or oropharyngeal tumors. Furthermore, MED15 expression correlated between primary tumors and corresponding lymph node metastases. MED15 correlated with proliferation in tissues, and MED15 knockdown reduced proliferation and migration. We observed an association between MED15 and TGF-β activity in tissues because TGF-β activation led to increased MED15 expression and reduced pSMAD3 on MED15 knockdown. Taken together, our results implicate MED15 in HNSCC and hint that MED15 overexpression is a clonal event during HNSCC progression. MED15 may serve as a prognostic marker for recurrence and as a therapeutic target.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture