Merck
Cerebrospinal fluid cortisol and clinical disease progression in MCI and dementia of Alzheimer's type.
Neurobiology of aging (2014-12-02)
Julius Popp, Steffen Wolfsgruber, Isabella Heuser, Oliver Peters, Michael Hüll, Johannes Schröder, Hans-Jürgen Möller, Piotr Lewczuk, Anja Schneider, Holger Jahn, Christian Luckhaus, Robert Perneczky, Lutz Frölich, Michael Wagner, Wolfgang Maier, Jens Wiltfang, Johannes Kornhuber, Frank Jessen
ABSTRAKT
Increased peripheral and central nervous system cortisol levels have been reported in Alzheimer's disease (AD) and may reflect dysfunction of cerebral components of the hypothalamic-pituitary-adrenal (HPA) axis. However, brain exposure to high cortisol concentrations may also accelerate disease progression and cognitive decline. The objectives of this study were to investigate whether HPA-axis dysregulation occurs at early clinical stages of AD and whether plasma and CSF cortisol levels are associated with clinical disease progression. Morning plasma and CSF cortisol concentrations were obtained from the subjects with AD dementia, mild cognitive impairment of AD type (MCI-AD), MCI of other type (MCI-O), and controls with normal cognition included in a multicenter study from the German Dementia Competence Network. A clinical and neuropsychological follow-up was performed in a subgroup of participants with MCI-AD, MCI-O, and AD dementia. CSF cortisol concentrations were increased in the subjects with AD dementia or MCI-AD compared with subjects with MCI-O or normal cognition. After controlling for possible confounders including CSF measures of amyloid beta1-42 and total tau, higher baseline CSF cortisol levels were associated with faster clinical worsening and cognitive decline in MCI-AD. The findings suggest that HPA-axis dysregulation occurs at the MCI stage of AD and may accelerate disease progression and cognitive decline.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Diphenylcyclopropenone, 98%
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard