Merck
An open-label phase Ib dose-escalation study of TRC105 (anti-endoglin antibody) with bevacizumab in patients with advanced cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-09-28)
Michael S Gordon, Francisco Robert, Daniela Matei, David S Mendelson, Jonathan W Goldman, E Gabriela Chiorean, Robert M Strother, Ben K Seon, William D Figg, Cody J Peer, Delia Alvarez, Bonne J Adams, Charles P Theuer, Lee S Rosen
ABSTRAKT
Endoglin, an endothelial cell membrane receptor expressed on angiogenic tumor vessels, is essential for angiogenesis and upregulated in the setting of VEGF inhibition. TRC105 is an anti-endoglin IgG1 monoclonal antibody that potentiates VEGF inhibitors in preclinical models. This study assessed safety, pharmacokinetics, and antitumor activity of TRC105 in combination with bevacizumab. Patients (n = 38) with advanced solid tumors, Eastern Cooperative Group performance status 0-1, and normal organ function were treated with escalating doses of TRC105 plus bevacizumab until disease progression or unacceptable toxicity using a standard 3 + 3 phase I design. TRC105 and bevacizumab were well tolerated at their recommended single-agent doses (10 mg/kg) when the initial dose of TRC105 was delayed by one week and divided over 2 days to limit the frequency of headache. The concurrent administration of bevacizumab and TRC105 did not otherwise potentiate known toxicities of TRC105 or bevacizumab. Hypertension and proteinuria were observed, though not at rates expected for single-agent bevacizumab. Several patients who had previously progressed on bevacizumab or VEGF receptor tyrosine kinase inhibitor (VEGFR TKI) treatment experienced reductions in tumor volume, including two partial responses by RECIST, and 6 remained without progression for longer periods than during their prior VEGF inhibitor therapy. TRC105 was well tolerated with bevacizumab and clinical activity was observed in a VEGF inhibitor-refractory population. Ongoing clinical trials are testing TRC105 in combination with bevacizumab in glioblastoma and with VEGFR TKIs in renal cell carcinoma, hepatocellular carcinoma, and soft tissue sarcoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Alanine, European Pharmacopoeia (EP) Reference Standard