Merck
HLA monomers as a tool to monitor indirect allorecognition.
Transplantation (2014-05-07)
Eytan Breman, Paula P van Miert, Dirk M van der Steen, Mirjam H Heemskerk, Ilias I Doxiadis, Dave Roelen, Frans H Claas, Cees van Kooten
ABSTRAKT
Recognition of donor antigens can occur through two separate pathways: the direct pathway (non-self HLA on donor cells) and the indirect pathway (self-restricted presentation of donor derived peptides on recipient cells). Indirect allorecognition is important in the development of humoral rejection; therefore, there is an increasing interest in the monitoring of indirect alloreactive T-cells. We have used an in vitro model to determine the optimal requirements for indirect presentation and assessed the risk for semidirect presentation in this system. HLA-typed monocyte-derived dendritic cells (moDCs) were incubated with cellular fragments or necrotic cells and incubated with either indirect or direct alloreactive T-cell clones. T-cell reactivity was measured through proliferation or cytokine secretion. HLA-typed moDC, monocytes, or PBMCs were incubated with HLA class I monomers, in combination with either direct/indirect T-cell clones. Although both were efficiently taken up, alloreactivity was limited to the semi-direct pathway, as measured by allospecific CD4 (indirect) and CD8 T-cell clones (direct) when cells were used. In contrast, HLA-A2 monomers were not only efficiently taken up but also processed and presented by HLA-typed moDC, monocytes, and PBMCs. Activation was shown by a dose-dependent induction of IFN-γ production and proliferation by the CD4 T-cell clone. Antigen presentation was most efficient when the monomers were cultured for longer periods (24-48 hr) in the presence of the T-cells. Using this method, no reactivity was observed by the CD8 T-cell clone, confirming no semidirect alloreactivity. We have developed a system that could be used to monitor indirect alloreactive T-cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
SAFC
L-Glutamine
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland