Merck
Lack of sexual privacy affects psychological and marital domains of male sexual dysfunction.
The journal of sexual medicine (2014-01-28)
Valentina Boddi, Alessandra Daphne Fisher, Elisa Maseroli, Giulia Rastrelli, Giovanni Corona, Emmanuele Jannini, Valdo Ricca, Edoardo Mannucci, Mario Maggi
ABSTRAKT
Sexual dysfunctions (SDs) are dictated by predisposing, precipitating, maintaining, and contextual factors, the latter of which can help sexual problems to emerge. Even if the lack of sexual privacy is one of the most common contextual issues, it has not been extensively studied. Investigation of sexual privacy in a large sample of men consulting for SD was the aim of this study. A consecutive series of 3,736 men, attending the outpatient clinic for SD for the first time, was retrospectively studied. Privacy during sexual intercourse was investigated with the following question, "During the last three months, have you had enough privacy during your sexual activity?" and rated 0 = yes, 1 = sometimes, 2 = rarely, and 3 = never. Several clinical, biochemical, and psychological (Middlesex Hospital Questionnaire [MHQ]) parameters were studied. Among the 3,736 patients studied, 83.9% reported enough privacy during sexual intercourse, while 8.6%, 5.7%, and 1.7% declared a decrease of sexual privacy of increasing severity. Lack of sexual privacy was associated with ejaculatory dysfunctions and with the inability to maintain an erection during intercourse. Subjects reporting lack of sexual privacy had a higher risk of relational and intrapsychic impairments, as well as psychopathology at MHQ questionnaire, even after adjusting for confounders. Fatherhood was associated with sexual privacy issues only in the lowest quartiles. In subjects without children, the absence of cohabitation with the partner was associated with an increasing risk of not having enough privacy (hazard ratio [HR] = 1.837 [1.269-2.659], P = 0.001); data confirmed, after stratification for age, only in the youngest subjects (I quartile HR = 2.159 [1.211-3.848], P = 0.009). This study indicates that sexual privacy is often a poorly investigated item, which is important to evaluate in male SD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications