Merck
  • Evolution of strategies to achieve baseline separation of ten anionic, water-soluble sulfated estrogens via achiral packed column supercritical fluid chromatography.
Evolution of strategies to achieve baseline separation of ten anionic, water-soluble sulfated estrogens via achiral packed column supercritical fluid chromatography.
Journal of chromatography. A (2014-12-03)
M A Patel, M A Hardink, L Wrisely, F W Riley, C J Hudalla, M Ashraf-Khorassani, L T Taylor
ABSTRAKT
Near baseline separation of ten sulfated sodium salts of various structurally related estrogens employing a variety of bonded stationary phase packed columns was obtained using a conventional supercritical fluid chromatograph coupled with UV detection. Critical pairs 2/3 (8,9-dehydroestrone/17β-dihydroequilin) and 6/7 (17α-estradiol or 17α-dihydroequilin/estrone), however, failed to baseline separate. In all preliminary separations, 10mM ammonium acetate and variable percentages of H2O were initially used as co-additives in conjunction with methanol as a modifier. Different modifier programs and temperatures were employed to optimize the separation in a timely manner. A 2-ethylpyridine column provided the best separation compared to bare silica, diol, and cyano-based bonded phase columns. The employment of both salt and water as additives to the methanol-modified CO2 mobile phase suggested a mixed mode separation mechanism involving both ion pairing of each anionic sulfated estrogen with ammonium ion and hydrophilic interaction facilitated by partitioning of analyte between the aqueous solvated stationary phase and the aqueous component of the mobile phase. Upon more extensive study with either iso-propylamine or formic acid-ammonium formate buffer, the critical anionic pairs were 95% baseline resolved.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Water, ACS reagent
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
Standardowa gęstość 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C