Merck
  • Mycological and aflatoxin contamination of peanuts sold at markets in Kinshasa, Democratic Republic of Congo, and Pretoria, South Africa.
Mycological and aflatoxin contamination of peanuts sold at markets in Kinshasa, Democratic Republic of Congo, and Pretoria, South Africa.
Food additives & contaminants. Part B, Surveillance (2014-06-11)
Ilunga Kamika, Pamella Mngqawa, John P Rheeder, Snow L Teffo, David R Katerere
ABSTRAKT
Peanut (Arachis hypogaea L.) is an important food crop in sub-Saharan Africa. In this survey, the mycological and aflatoxin contamination of peanuts collected from Kinshasa, Democratic Republic of Congo, and Pretoria, South Africa, was assessed. Twenty peanut samples were purchased randomly at informal markets in the two cities and analysed for mycoflora and aflatoxins (AFB1, AFB2, AFG1 and AFG2) using standard methods. The results indicated that 95% of the Kinshasa samples and 100% of the Pretoria samples were contaminated with aflatoxigenic fungi in the ranges 20-49,000 and 40-21,000 CFU/g, respectively. Seventy-five per cent of the Kinshasa samples and 35% of the Pretoria samples exceeded the maximum limits of AFB1 as set by The Joint FAO/WHO Expert Committee on Food Additives. Residents of both cities are at a high risk of aflatoxin exposure despite their apparent cultural, socio-economic, geographic and climatic differences. Further work needs to be done to understand the supply chains of peanut trade in informal markets of the two countries so that interventions are well targeted on a regional rather than a national level.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material