Merck
Preparation and pharmaceutical evaluation of new tacrolimus-loaded solid self-emulsifying drug delivery system.
Archives of pharmacal research (2014-08-20)
Youn Gee Seo, Dong-Wuk Kim, Kwan Hyung Cho, Abid Mehmood Yousaf, Dong Shik Kim, Jeong Hoon Kim, Jong Oh Kim, Chul Soon Yong, Han-Gon Choi
ABSTRAKT
The purpose of this study was to develop a novel tacrolimus-loaded solid self-emulsifying drug delivery system (SEDDS) using Labrafac as an oil phase. The ternary phase diagram was plotted with Labrafac, Labrasol and Lauroglycol used as an oil, surfactant and co-surfactant, respectively. The liquid SEDDS formulated with Labrasol, Lauroglycol and Labrafac (70:15:15, volume ratio) furnished the smallest emulsion globule size. The solid SEDDS was obtained by spray-drying the liquid mixture containing the liquid SEDDS with 5 % tacrolimus and silicon dioxide. Furthermore, dissolution of tacrolimus from the solid SEDDS and pharmacokinetics in rats was studied compared to the commercial product. The solid SEDDS produced relatively larger emulsion globule size than that exhibited by the corresponding liquid SEDDS. However, this size variation was not significantly different. The solid SEDDS with approximately 280 nm emulsion droplet size improved the dissolution of the drug compared to drug power and the commercial product. It resulted in significantly higher plasma concentration, AUC and Cmax, and shorter Tmax values than did the commercial product (p < 0.05). The enormously enhanced oral bioavailability of tacrolimus in rats was attributed to relatively faster absorption due to accelerated dissolution of the drug from the solid SEDDS. Therefore, this novel solid SEDDS prepared with Labrafac as an oil phase is an excellent way to achieve better bioavailability of tacrolimus given via the oral route.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Diethyl ether
Supelco
Diethyl ether, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free