Merck
Diagnosis and treatment of low-grade osteosarcoma: experience with nine cases.
International journal of clinical oncology (2013-07-25)
Katsuhiro Hayashi, Hiroyuki Tsuchiya, Norio Yamamoto, Toshiharu Shirai, Hideji Nishida, Akihiko Takeuchi, Hiroaki Kimura, Shinji Miwa, Hiroyuki Inatani, Hideki Okamoto, Satoshi Yamada, Hiroko Ikeda, Seiko Sawada-Kitamura, Takayuki Nojima, Akishi Ooi, Takanobu Otsuka
ABSTRAKT
Low-grade osteosarcoma, including low-grade central osteosarcoma and parosteal osteosarcoma, is an extremely rare variant, and the diagnosis is occasionally difficult. In this article we present cases of low-grade osteosarcomas that should be reviewed by a clinical oncologist. Nine cases of histologically diagnosed Broder grade 1 osteosarcoma were retrospectively reviewed. The pathological diagnoses included parosteal osteosarcoma, low-grade central osteosarcoma, and low-grade chondroblastic osteosarcoma in four, four, and one cases, respectively. Duration from initial surgical intervention including biopsy to final diagnosis as low-grade osteosarcoma was a mean of 9.4 months. The initial benign diagnoses on biopsy specimens included fibrous dysplasia in three cases, chondroblastoma in one case, and a giant cell tumor in one case. The average number of histological examinations was 1.8. Low-grade osteosarcomas are well suited for biological reconstruction: seven cases were reconstructed by frozen autografts, distraction osteogenesis, or vascularized bone grafts. Low-grade osteosarcomas can be misdiagnosed as benign lesions, especially fibrous dysplasia. If the diagnosis of a low-grade osteosarcoma is not established on the basis of radiologic findings, care should be exercised, even when a biopsy suggests a benign lesion. Low-grade osteosarcomas should be treated with wide excision, even after an intralesional excision. Biological reconstruction might be a better option for low-grade osteosarcomas.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%