Merck
Haemoconcentration of residual cardiopulmonary bypass blood using Hemosep ®: a randomised controlled trial.
Anaesthesia (2015-02-19)
M Hogan, A Needham, E Ortmann, F Bottrill, T J Collier, M W Besser, A A Klein
ABSTRAKT
Cardiac surgery and cardiopulmonary bypass are associated with haemodilution, activation of haemostasis and blood transfusion. We undertook a randomised controlled trial that included 53 patients in order to compare autotransfusion of residual cardiopulmonary bypass blood with residual blood concentrated using the novel Hemosep(®) device. There was no difference in patients' mean (SD) haemoglobin concentration after autotransfusion of unprocessed blood compared with Hemosep; 103.5 (10.2) g.l(-1) vs 106.2 (12.4) g.l(-1), respectively, p = 0.40. The mean (SD) change in haemoglobin concentration after autotransfusion was 5.9 (5.3) g.l(-1) in the control group compared with 4.9 (6.3) g.l(-1) in the Hemosep group, p = 0.545. Adjusted for baseline haemoglobin concentrations, the estimated mean (95% CI) difference in change in haemoglobin concentration (control vs Hemosep) was 0.57 (-2.65 to 3.79) g.l(-1), p = 0.72. This was despite Hemosep's reducing the weight of the blood from a mean (SD) of 778.7 (243.0) g to 607.3 (248.2) g, p < 0.001. The haemoglobin concentration in the processed blood increased from a mean (SD) of 87.0 (15.1) g.l(-1) to 103.7 (17.4) g.l(-1), p < 0.001. We conclude that Hemosep is capable of haemoconcentration when employed to process residual cardiopulmonary bypass blood, but that this is insufficient to increase patient haemoglobin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Mannitol, European Pharmacopoeia (EP) Reference Standard