Merck
Effect of alendronate sodium on tooth movement in ovariectomized rats.
Archives of oral biology (2015-03-15)
Marcio Salazar, Luzmarina Hernandes, Adilson Luiz Ramos, Brunamélia de Oliveira Salazar, Kelly Regina Micheletti, Luiz Renato Paranhos, Marcos Rogério de Mendonça, Osmar Aparecido Cuoghi
ABSTRAKT
The aim of the present study was to evaluate the effect of two different dosages of alendronate on induced orthodontic movement in an experimental model involving rats with osteoporosis following ovariectomy. Female Wistar rats (Rattus norvegicus) eight weeks of age were divided into four groups (n=12/group): ovariectomized (OVX group); ovariectomized and treated with alendronate sodium at 1mg/kg (Group OVX+ALN1); ovariectomized and treated with alendronate sodium at 2mg/kg (Group OVX+ALN2); and sham operated (control). Three months after ovariectomy, the maxillary right first molar was submitted to movement for five and seven days. After the death of the animals, the maxilla were removed and processed for microscopic evaluation. The maxillary left first molar (without movement) was used for comparison purposes in all groups. The samples were processed for the quantification of alveolar bone and tooth movement. Intragroup comparisons showed significant movement after five and seven days (p<0.05) for all groups. Comparison among groups revealed greater tooth movement in the OVX group (p<0.05), on day 7. Both alendronate sodium doses similarly decreased tooth movement in ovariectomized rats (p>0.05). Movement in ovariectomized+alendronate groups were also smaller than non-ovariectomized rats, however without statistical difference.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alendronate sodium trihydrate, ≥97% (NMR), powder
USP
Alendronian sodu, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Supelco
Diethylene glycol, analytical standard
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard