Merck
Membrane fouling by vesicles and prevention through ozonation.
Environmental science & technology (2014-06-10)
Michelle C Barry, Kiril Hristovski, Paul Westerhoff
ABSTRAKT
Membrane fouling is a major challenge in water and wastewater treatment. Recent observations that ozone mitigates membrane fouling during filtration of secondary effluent prompted this study into the impact of preozonation on membrane fouling caused by biogenic colloids. The focus of this study was on liposomes, synthetic vesicles composed of (phospho)lipid bilayers, which are representative of the diverse cellular vesicles present in all biologically impacted waters. The overarching hypothesis was that these biologically produced, nonrigid or "soft" colloids (e.g., vesicles) present in wastewater give rise to unique fouling behavior that can be mitigated by preozonation. Using dead-end ultrafiltration (UF) and batch ozonation tests, the key findings of this study were (1) liposomes fouled UF membranes faster (4-13 times membrane cake resistance (RC) per mgC filtered) than polysaccharides, fatty acids, and NOM on a DOC-normalized basis; (2) based on the estimated carbon distribution of secondary effluent, liposome-like biogenic nanomaterials could be responsible for 20-60% of fouling during UF; and (3) preozonation reduces liposomal fouling during UF, likely due to the disruption of the liposome structure through cleavage of the fatty acid tails at carbon-carbon double bonds.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Supelco
Caprylic Acid (Octanoic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Caprylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Supelco
Octanoic acid, analytical standard