Merck
  • The mitochondrial-targeted antioxidant MitoQ ameliorates metabolic syndrome features in obesogenic diet-fed rats better than Apocynin or Allopurinol.
The mitochondrial-targeted antioxidant MitoQ ameliorates metabolic syndrome features in obesogenic diet-fed rats better than Apocynin or Allopurinol.
Free radical research (2014-07-30)
Christine Feillet-Coudray, Gillen Fouret, Raymond Ebabe Elle, Jennifer Rieusset, Beatrice Bonafos, Beatrice Chabi, David Crouzier, Kamelija Zarkovic, Neven Zarkovic, Jeanne Ramos, Eric Badia, Michael P Murphy, Jean Paul Cristol, Charles Coudray
ABSTRAKT
The prevalence of metabolic syndrome (MetS) components including obesity, dyslipidemia, insulin resistance (IR), and hepatic steatosis is rapidly increasing in wealthy societies. It is accepted that inflammation/oxidative stress are involved in the initiation/evolution of the MetS features. The present work was designed to evaluate the effects of three major cellular ROS production systems on obesity, glucose tolerance, and hepatic steatosis development and on oxidative stress onset. To do so, 40 young male Sprague-Dawley rats were divided into 5 groups: 1-control group, 2-high fat (HF) group (60% energy from fat), 3-HF+ MitoQ (mitochondrial ROS scavenger), 4-HF+ Apocynin (NADPH oxidase inhibitor), 5-HF+ Allopurinol (xanthine oxidase inhibitor). After 8 weeks of these treatments, surrogate MetS, mitochondrial function, and oxidative stress markers were measured in blood and liver. As expected, rats that were fed the HF diet exhibited increased body weight, glucose intolerance, overt hepatic steatosis, and increased hepatic oxidative stress. The impacts of the studied ROS inhibitors on these aspects of the MetS were markedly different. MitoQ showed the most clinically relevant effects, attenuating body weight gain and glucose intolerance provoked by the HF diet. Both Apocynin and Allopurinol showed limited effects suggesting secondary roles of xanthine oxidase (XO) or NADPH oxidase-dependent ROS production in the onset of oxidative stress-dependent obesity, glucose intolerance, and hepatic steatosis process. Thus, MitoQ revealed the central role of mitochondrial oxidative stress in the development of MetS and suggested that mitochondria-targeted antioxidants may be worth considering as potentially helpful therapies for MetS features.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard