Merck
Effects of sex hormones on inflammatory response in male and female vascular endothelial cells.
Journal of endocrinological investigation (2014-06-21)
Giosuè Annibalini, Deborah Agostini, Cinzia Calcabrini, Chiara Martinelli, Evelin Colombo, Michele Guescini, Pasquale Tibollo, Vilberto Stocchi, Piero Sestili
ABSTRAKT
Gender-related differences in sex hormones might have a key role in the development of atherosclerosis though direct vascular effects of sex hormones are not yet well understood. Thus, the main purpose of this study was to compare the effects of sex hormones on inflammatory response in Human Umbilical Vein Endothelial Cells (HUVECs) obtained from both male and female donors. We analyzed the expression of receptors and enzymes relevant to the action of androgens (AR, 5α-reductase 1 and 5α-reductase 2) and estrogens (ERα, ERβ, and aromatase) in male and female HUVECs. Furthermore, we analyzed the effect of testosterone (T), 17β-estradiol (E2), dihydrotestosterone (DHT), and several androgenic-anabolic steroids (AAS) on VCAM-1, ICAM-1, and E-selectin gene expression and on adhesion of U937 cells to TNF-α-stimulated male and female HUVECs. Our results reveal that in HUVECs, regardless of gender, the components involved in the androgen action pathway are predominant as compared to those of estrogen action pathway. In both HUVEC genders, the inflammatory effect of TNF-α was amplified by co-administration of T or DHT and several AAS frequently used in doping, while E2 had no effect. This is the first study analyzing, under identical culture conditions, the key components of sex hormone response in male and female HUVECs and the possible role of sex hormones in regulating the endothelial inflammatory response. The data obtained in our experimental system showed a pro-inflammatory effect of androgens, while conclusively excluding any protective effect for all the tested hormones.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution