Merck
Microbe-specific unconventional T cells induce human neutrophil differentiation into antigen cross-presenting cells.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-29)
Martin S Davey, Matt P Morgan, Anna Rita Liuzzi, Christopher J Tyler, Mohd Wajid A Khan, Tamas Szakmany, Judith E Hall, Bernhard Moser, Matthias Eberl
ABSTRAKT
The early immune response to microbes is dominated by the recruitment of neutrophils whose primary function is to clear invading pathogens. However, there is emerging evidence that neutrophils play additional effector and regulatory roles. The present study demonstrates that human neutrophils assume Ag cross-presenting functions and suggests a plausible scenario for the local generation of APC-like neutrophils through the mobilization of unconventional T cells in response to microbial metabolites. Vγ9/Vδ2 T cells and mucosal-associated invariant T cells are abundant in blood, inflamed tissues, and mucosal barriers. In this study, both human cell types responded rapidly to neutrophils after phagocytosis of Gram-positive and Gram-negative bacteria producing the corresponding ligands, and in turn mediated the differentiation of neutrophils into APCs for both CD4(+) and CD8(+) T cells through secretion of GM-CSF, IFN-γ, and TNF-α. In patients with acute sepsis, circulating neutrophils displayed a similar APC-like phenotype and readily processed soluble proteins for cross-presentation of antigenic peptides to CD8(+) T cells, at a time when peripheral Vγ9/Vδ2 T cells were highly activated. Our findings indicate that unconventional T cells represent key controllers of neutrophil-driven innate and adaptive responses to a broad range of pathogens.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
IFN-γ human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture