Merck
Spirocerca lupi infection in a dog from southern Italy: an "old fashioned" disease?
Parasitology research (2014-05-02)
Alessio Giannelli, Valeria Baldassarre, Rafael A N Ramos, Riccardo P Lia, Tommaso Furlanello, Michele Trotta, Filipe Dantas-Torres, Gad Baneth, Domenico Otranto
ABSTRAKT
Canine spirocercosis caused by Spirocerca lupi is a life-threatening helminthic disease featured by severe clinical signs and potential development of oesophageal neoplasia. This infection is considered fairly frequent in Europe but almost unknown in Italy, from where only few reports have been published in local journals at the beginning of the XXI century. In the present study, an autochthonous case of canine spirocercosis in a 2-year-old dog from southern Italy is described. The animal was admitted to a private veterinary clinic in the municipality of Potenza (Basilicata region) due to persistent dyspnoea, vomiting, and regurgitation. At the abdominal ultrasound, a mass (1.3 × 2 cm) was observed and, thereafter, surgically removed from the apical part of the stomach. A female specimen of S. lupi was morphologically identified during the histological examination of the nodule and its eggs were detected in the faeces. In addition, the morphological identification was confirmed by molecular amplification and sequencing of partial cox2 gene sequence of S. lupi. Veterinarians should be aware of the potential risks derived from infection with this spirurid, which requires specific diagnostic and preventive measures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Supelco
Prednisolone, VETRANAL®, analytical standard
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Prednisolone, ≥99%
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
Prednisolone, British Pharmacopoeia (BP) Assay Standard
USP
Prednisolone, United States Pharmacopeia (USP) Reference Standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard