Merck
Mullinamides A and B, new cyclopeptides produced by the Ruth Mullins coal mine fire isolate Streptomyces sp. RM-27-46.
The Journal of antibiotics (2014-04-10)
Xiachang Wang, Khaled A Shaaban, Sherif I Elshahawi, Larissa V Ponomareva, Manjula Sunkara, Gregory C Copley, James C Hower, Andrew J Morris, Madan K Kharel, Jon S Thorson
ABSTRAKT
Two new cyclopeptides, mullinamides A [cyclo-(-L-Gly-L-Glu-L-Val-L-Ile-L-Pro-)] and B [cyclo-(-L-Glu-L-Met-L-Pro-)] were isolated from the crude extract of terrestrial Streptomyces sp. RM-27-46 along with the three known cyclopeptides surugamide A [cyclo-(-L-Ile-D-Ile-L-Lys-L-Ile-D-Phe-D-Leu-L-Ile-D-Ala-)], cyclo-(-L-Pro-L-Phe-) and cyclo-(-L-Pro-L-Leu-). The structures of the new compounds were elucidated by the cumulative analyses of NMR spectroscopy and HRMS. Although mullinamides A and B displayed no appreciable antimicrobial/fungal activity or cytotoxicity, this study highlights the first reported antibacterial activity of surugamide A.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material