Merck
  • Composite chitosan/silk fibroin nanofibers for modulation of osteogenic differentiation and proliferation of human mesenchymal stem cells.
Composite chitosan/silk fibroin nanofibers for modulation of osteogenic differentiation and proliferation of human mesenchymal stem cells.
Carbohydrate polymers (2014-07-20)
Guo-Jyun Lai, K T Shalumon, Shih-Hsien Chen, Jyh-Ping Chen
ABSTRAKT
Nanofibrous membrane scaffolds of chitosan (CS), silk fibroin (SF) and CS/SF blend were prepared by electrospinning and studied for growth and osteogenic differentiation of human bone marrow mesenchymal stem cells (hMSCs). The morphology and physico-chemical properties of all membrane scaffolds were compared. The influence of CS and SF on cell proliferation was assessed by the MTS assay, whereas osteogenic differentiation was determined from the Alizarin Red staining, alkaline phosphatase activity and expression of osteogenic marker genes. The osteogenic differentiation and proliferation of hMSCs were enhanced by CS and SF nanofibers, respectively. Blending CS with SF retained the osteogenesis nature of CS without negatively influencing the cell proliferative effect of SF. By taking advantage of the differentiation/proliferation cues from individual components, the electrospun CS/SF composite nanofibrous membrane scaffold is suitable for bone tissue engineering.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Cetylpyridinium chloride, European Pharmacopoeia (EP) Reference Standard
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
USP
Chlorek cetylopirydyniowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer