Merck
Rheinheimera japonica sp. nov., a novel bacterium with antimicrobial activity from seashore sediments of the Sea of Japan.
Archives of microbiology (2015-02-24)
L A Romanenko, N Tanaka, V I Svetashev, N I Kalinovskaya, V V Mikhailov
ABSTRAKT
Two gram-negative, aerobic, brown-pigmented, motile rod-shaped bacteria KMM 9512 and KMM 9513(T) were isolated from a sediment sample collected from the Sea of Japan seashore, Russia. On the basis of the 16S rRNA gene sequence analysis, novel strains KMM 9512 and KMM 9513(T) positioned within the genus Rheinheimera (class Gammaproteobacteria) as a separate subline adjacent to Rheinheimera baltica DSM 14885(T) sharing highest gene sequence similarities of 98.6-97.6 % to their closest phylogenetic relatives, Rheinheimera muenzenbergensis LMG 27269(T), R. baltica DSM 14885(T), Rheinheimera aquimaris JCM 14331(T), Rheinheimera nanhaiensis KACC 14030(T), and Rheinheimera pacifica KMM 1406(T). Strains KMM 9512 and KMM 9513(T) belong to the same separate genospecies on the basis of the 16S rRNA gene sequence similarity (99.8 %) and their DNA relatedness to each other (89 %) and to closely related Rheinheimera species (25-53 %). The major isoprenoid quinone was Q-8, polar lipids consisted of phosphatidylserine, phosphatidylglycerol, phosphatidylethanolamine, unknown aminolipids, unknown aminophospholipids, unknown phospholipids, and unknown lipids, and major fatty acid were C16:0, C16:1 ω7c, C17:1 ω8c, C12:0 3-OH followed by C17:0 and C18:1 ω7c in both strains. Strains KMM 9512 and KMM 9513(T) revealed a remarkable antagonistic activity toward a number of gram-positive and gram-negative microorganisms. On the basis of phylogenetic analysis, DNA-DNA hybridization results, and phenotypic differences, strains KMM 9512 and KMM 9513(T) are proposed to be classified as a novel species of the genus Rheinheimera, Rheinheimera japonica sp. nov. The type strain of this species is KMM 9513(T) = NRIC 0918(T).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%