Merck
  • High-performance liquid chromatographic enantioseparation of cationic 1,2,3,4-tetrahydroisoquinoline analogs on Cinchona alkaloid-based zwitterionic chiral stationary phases.
High-performance liquid chromatographic enantioseparation of cationic 1,2,3,4-tetrahydroisoquinoline analogs on Cinchona alkaloid-based zwitterionic chiral stationary phases.
Analytical and bioanalytical chemistry (2014-10-20)
István Ilisz, Nóra Grecsó, Ferenc Fülöp, Wolfgang Lindner, Antal Péter
ABSTRAKT
The stereoisomers of 1,2,3,4-tetrahydroisoquinoline analogs were resolved for the first time by applying a polar ionic mobile phase on a quinine or a quinidine moiety fused with a chiral sulfonic acid-type chiral selector immobilized on silica [Chiralpak ZWIX(+)™ and Chiralpak ZWIX(-)™]. The effects of the nature and concentrations of the mobile phase components and additives and temperature on the retention and enantioseparation on the investigated chiral columns were studied. Experiments were performed in the temperature range 10-50 °C. Thermodynamic parameters were calculated from plots of ln α versus 1/T. The separations were generally enthalpy-controlled, but entropy-controlled separation was also observed below 30 °C. The enantiomer elution order was determined in some cases and was observed to be opposite on the ZWIX(+)™ and ZWIX(-)™ columns. Our results contribute to a better understanding of the enantiorecognition mechanism of chiral bases with chiral zwitterionic selectors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%