Merck
Unbiased modifier screen reveals that signal strength determines the regulatory role murine TLR9 plays in autoantibody production.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-15)
Robyn E Mills, Viola C Lam, Allison Tan, Nicole Cresalia, Nir Oksenberg, Julie Zikherman, Mark Anderson, Arthur Weiss, Michelle L Hermiston
ABSTRAKT
The autoimmune disease systemic lupus erythematosus has a complex environmental and multifactorial genetic basis. Genome-wide association studies have recently identified numerous disease-associated polymorphisms, but it remains unclear in which cells and during which step of pathogenesis specific polymorphisms interact to cause disease. Using a mouse model in which the same activating mutation (CD45E613R) causes distinct genetic background-dependent disease phenotypes, we performed a screen for genetic modifiers of autoreactivity between anti-nuclear Ab (ANA)-resistant CD45E613R.B6 and ANA-permissive CD45E613R.BALB/c mice. Within a novel autoreactivity-associated locus on chromosome 9, we identify a putative modifier, TLR9. Validating a role for TLR9 in modifying autoreactivity in the context of the CD45E613R mutation, manipulation of TLR9 gene dosage eliminates ANA in CD45E613R.BALB/c mice, but confoundingly permits ANA in CD45E613R.B6 mice. We demonstrate that sensitivity to ANA is modulated by strength of TLR9 signal, because stronger TLR9(B6) signals, but not weaker TLR9(BALB/c) signals, negatively regulate CD45E613R B cell development during competitive reconstitution at the central tolerance checkpoint. Our results identify a novel autoreactivity-associated locus and validate Tlr9 as a candidate gene within the locus. We further demonstrate a novel role for TLR9 signal strength in central tolerance, providing insight into the interplay of disease-associated polymorphisms at a discrete step of systemic lupus erythematosus pathogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Poly(deoxyadenylic-thymidylic) acid sodium salt, double-stranded alternating copolymer
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard