Merck
  • Determination of adsorption affinity of nanoparticles for interleukin-8 secreted from A549 cells by in vitro cell-free and cell-based assays.
Determination of adsorption affinity of nanoparticles for interleukin-8 secreted from A549 cells by in vitro cell-free and cell-based assays.
Journal of toxicology and environmental health. Part A (2014-12-17)
Yun-Geon Lee, Jiyoung Jeong, Jennifer Raftis, Wan-Seob Cho
ABSTRAKT
The evaluation of the potential of nanoparticles (NP) for adsorbing biomolecules and use of control approaches are important for accurate presentation of in vitro analytical data. In this study, seven types of NP including carbon black (CB), cerium dioxide (CeO2), copper oxide (CuO), indium trioxide (In2O3), nickel oxide (NiO), silicon dioxide (SiO2), and titanium dioxide (TiO2) were used for determining the adsorption ability of interleukin-8 (IL-8) under either (1) a cell-free condition where NP were incubated with supernatant of A549 cells, or (2) a cell-based condition, where cells were treated with NP. Under the cell-free condition, CB and TiO2 NP showed a high adsorption affinity for IL-8 in supernatants of both lipopolysaccharide (LPS)-stimulated and unstimulated A549 cells. In contrast, SiO2 and In2O3 NP displayed a relatively low adsorption affinity. Further, IL-8 adsorption was markedly reduced when NP were predispersed in fetal bovine serum. The results obtained under cell-based conditions using both stimulated and unstimulated cells were consistent with those of the cell-free condition. Data indicate that adsorption of IL-8 onto NP surface is variable depending on type of NP, preparation method of NP, and cellular inflammatory state. Thus, the cell-free adsorption assay may be utilized for reliable interpretation of data produced by in vitro cell-based methodology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, nanowires, diam. × L ~20 nm × 10 μm
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
SAFC
L-Glutamine
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%