Merck
Chemical composition and antioxidant activity of ultrasound-assisted extract of the endemic plant Haberlea rhodopensis Friv.
Journal of food science and technology (2015-04-02)
Dasha Mihaylova, Mirena Ivanova, Slava Bahchevanska, Albert Krastanov
ABSTRAKT
The antioxidant capacity of extracts from leaves of Haberlea rhodopensis Friv. obtained by a highly efficient and simple method of the ultrasound-assisted extraction was investigated. The total polyphenolic content was determined spectrophotometrically using the Folin - Ciocalteu's phenol reagent and the polyphenols composition was analyzed by HPLC method. Antioxidant activity of the extract was evaluated as 2,2-diphenyl-1-picrylhydrazyl (DPPH) radical scavenging activity, 2,2-azinobis-3 ethyl benxothiazoline-6-sulphonic acid (ABTS) cation decolorization activity assays and the ferric reducing/antioxidant power method (FRAP). The total phenolics of the extract were determined to be 148.71 ± 5.33 mg GAE on a dry weight basis. The HPLC analysis showed a large number of flavonoids and phenolic acids present in the ultrasound ethanol extract of H. rhodopensis. Results also revealed that the major flavonoids were luteolin (2730.18 μg/g DW), hesperidin (928.56 μg/g DW) and kaempferol (578.52 μg/g DW), whereas the most abundant phenolic acids identified in the extract were ferulic acid (630.48 μg/g DW) and sinapic acid (580.80 μg/g DW). DPPH(•) and ABTS(•+) values were 0.803 ± 0.007 and 0.769 ± 0.040 mmol TE/g DW, respectively and FRAP values were 1.362 ± 0.05 mmol TE/g DW.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%