Merck
A moldable putty containing silk fibroin yolk shell particles for improved hemostasis and bone repair.
Advanced healthcare materials (2014-10-10)
Kushagra Saran, Pujiang Shi, Shashi Ranjan, James C H Goh, Yong Zhang
ABSTRAKT
During minimally invasive orthopedic surgeries, surgical intervention is required at two stages; to attain hemostasis and subsequently to implant the bone graft or its substitute. There is an apparent need for a material which can simultaneously control bone bleeding and provide support for bone repair. In this work, a moldable putty, which can be applied to bone defects (usually irregular in shape), was developed to address this need. It comprises of a hemostatic factor thrombin, osteoinductive "yolk-shell" particles containing bone growth factor (BMP-2), and an osteoconductive component hydroxyapatite. The yolk shell particles allowed controlled release of BMP-2 and showed significantly enhanced osteogenic differentiation of C2C12 (mouse myoblast) cells as demonstrated by increased alkaline phosphatase (ALP) activity and relative gene expressions of osteogenic differentiation markers. These particles were assembled into a moldable putty by mixing them with hydroxyapatite and silk fibroin solution (binding agent) supplemented with thrombin. The putty showed non-cytotoxicity, hemostatic ability, sustained release of BMP-2 and induced increased mineralization in C2C12 cells. This putty, if applied to bone defects during surgeries, may help attain hemostasis and may enhance bone repair by providing sustained release of bone growth factors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis