Merck
  • A novel adipocytokine, omentin, inhibits platelet-derived growth factor-BB-induced vascular smooth muscle cell migration through antioxidative mechanism.
A novel adipocytokine, omentin, inhibits platelet-derived growth factor-BB-induced vascular smooth muscle cell migration through antioxidative mechanism.
American journal of physiology. Heart and circulatory physiology (2014-04-15)
Kyosuke Kazama, Muneyoshi Okada, Hideyuki Yamawaki
ABSTRAKT
Omentin is a novel adipocytokine expressed in visceral adipose tissue. Secretion and blood concentration of omentin decrease in the obese subjects. We previously demonstrated that omentin is anti-inflammatory in vascular smooth muscle cells (SMCs). While vascular remodeling via migration of SMCs is also important for hypertension development, it remains to be clarified whether omentin affects this process. Here we examined whether omentin controls SMC migration. Omentin (300 ng/ml, 2 h) significantly inhibited platelet-derived growth factor (PDGF)-BB (10 ng/ml, 6 h)-induced migration of rat mesenteric arterial SMCs, as determined by Boyden chamber assay. Omentin (300 ng/ml, 2 h) significantly inhibited PDGF-BB (10 ng/ml, 30 min)-induced phosphorylation of p38 and heat shock protein (HSP) 27. Omentin (300 ng/ml, 2 h) significantly inhibited PDGF-BB (10 ng/ml, 30 min)-induced NADPH oxidase (NOX) activation as determined by lucigenin assay. Omentin (300 ng/ml, 24 h) significantly inhibited fetal bovine serum (5%, 4 days)-induced SMC outgrowth from rat isolated mesenteric artery. In vivo, omentin significantly inhibited carotid intimal hyperplasia in mouse ligation model. In summary, we for the first time demonstrate that omentin prevents PDGF-BB-induced SMC migration by preventing NOX/O2(-)/p38/HSP27 pathways, which might be at least partly responsible for the preventive effects on neointimal hyperplasia. Our data suggest that omentin may be protective against hypertension development by inhibiting vascular structural remodeling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard