Merck
  • In vitro - in vivo evaluation of a new oral dosage form of tramadol hydrochloride--controlled-release capsules filled with coated pellets.
In vitro - in vivo evaluation of a new oral dosage form of tramadol hydrochloride--controlled-release capsules filled with coated pellets.
Acta poloniae pharmaceutica (2014-10-01)
Danuta Szkutnik-Fiedler, Monika Balcerkiewicz, Wiesław Sawicki, Tomasz Grabowski, Edmund Grześkowiak, Jarosław Mazgalski, Hanna Urjasz
ABSTRAKT
The aim of this study was an in vitro - in vivo evaluation of a new oral dosage form of tramadol hydrochloride (TH), controlled-release capsules filled with coated pellets, 100 mg (TC), compared to the sustained release tablets, Tramal Retard, 100 mg (TR). In vitro release study of both formulations showed a similar release profile of TH over 8 h (f2 was 52). In vivo study (single oral, 100 mg dose administration in 8 rabbits) showed that the amount of TH absorbed into the systemic circulation after TC and TR administration was also similar (90% CI for AUC(0-t) and AUC(0-infinity) were 90-124% and 97-109%, respectively). However, a comparison of AUC(0-t) of pharmacokinetics of TC and TR indicates significantly prolonged absorption and elimination processes of TH when the drug is given in controlled-release capsules filled with coated pellets. It was manifested by longer: mean absorption time (p = 0.0016), mean residence time (p = 0.0268), absorption half-life (p = 0.0016), elimination half-life (p = 0.0493) and lower: absorption rate constant (p = 0.0016), elimination rate constant (p = 0.0148) and total body clearance Cl/F (p = 0.0076). It may be concluded that the new TH formulation could be expected to have a more prolonged analgesic activity than commercial sustained release tablets.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Tramadol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%