Merck
Evaluation of antioxidant activities of ampelopsin and its protective effect in lipopolysaccharide-induced oxidative stress piglets.
PloS one (2014-10-01)
Xiang Hou, Jingfei Zhang, Hussain Ahmad, Hao Zhang, Ziwei Xu, Tian Wang
ABSTRAKT
The aim of this study was to investigate the antioxidant potential of ampelopsin (APS) by using various methods in vitro, as well as to determine effects of APS on LPS-induced oxidative stress in piglets. The results showed that APS exhibited excellent free radical scavenging by DPPH, ABTS, O2•-, H2O2 and ferric reducing antioxidant power. Ampelopsin also protected pig erythrocytes against AAPH-induced apoptosis and hemolysis, decreased total superoxide dismutase activity, and increased lipid peroxidation. Furthermore the results demonstrated that APS enhanced the total antioxidant capacity and decreased the malondialdehyde and protein carbonyl contents in LPS-treated piglets. The results of the present investigation suggest that APS possesses a strong antioxidant activity and alleviates LPS-induced oxidative stress, possibly due to its ability to prevent reactive oxygen species.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%