Merck
Role of blood and vascular smooth muscle in the vasoactivity of nitrite.
American journal of physiology. Heart and circulatory physiology (2014-08-12)
Taiming Liu, Hobe J Schroeder, Lisa Barcelo, Shannon L Bragg, Michael H Terry, Sean M Wilson, Gordon G Power, Arlin B Blood
ABSTRAKT
Recent evidence from humans and rats indicates that nitrite is a vasodilator under hypoxic conditions by reacting with metal-containing proteins to produce nitric oxide (NO). We tested the hypothesis that near-physiological concentrations of nitrite would produce vasodilation in a hypoxia- and concentration-dependent manner in the hind limb of sheep. Anesthetized sheep were instrumented to measure arterial blood pressure and femoral blood flows continuously in both hind limbs. Nitrite was infused into one femoral artery to raise the nitrite concentration in the femoral vein by 10 to 15-fold while the sheep breathed 50%, 14% or 12% oxygen in inspired air. In contrast to reports in humans and rats, the nitrite infusion had no measurable effect on mean femoral blood flows or vascular conductances, regardless of inspired O2 levels. In vitro experiments showed no significant difference in the release of NO from nitrite in sheep and human red blood cells. Further experiments demonstrated nitrite is converted to NO in rat artery homogenates faster than sheep arteries, and that this source of NO production is attenuated in the presence of a heme oxidizer. Finally, western blots indicate that concentrations of the heme-containing protein cytoglobin, but not myoglobin, are markedly lower in sheep arteries compared with rats. Overall, the results demonstrate that nitrite is not a physiological vasodilator in sheep. This is likely due to a lack of conversion of nitrite to NO within the vascular smooth muscle, perhaps due to deficient amounts of the heme-containing protein cytoglobin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Pentobarbital, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexamethonium bromide