Merck
Pancreatic ductal perfusion at organ procurement enhances islet yield in human islet isolation.
Pancreas (2014-07-25)
Morihito Takita, Takeshi Itoh, Masayuki Shimoda, Mazhar A Kanak, Rauf Shahbazov, Faisal Kunnathodi, Michael C Lawrence, Bashoo Naziruddin, Marlon F Levy
ABSTRAKT
Pancreas preservation is a major factor influencing the results of islet cell transplantation. This study evaluated the effects of 2 different solutions for pancreatic ductal perfusion (PDP) at organ procurement. Eighteen human pancreases were assigned to 3 groups: non-PDP (control), PDP with ET-Kyoto solution, and PDP with cold storage/purification stock solution. Pancreatic islets were isolated according to the modified Ricordi method. No significant differences in donor characteristics, including cold ischemia time, were observed between the 3 groups. All islet isolations in the PDP groups had more than 400,000 islet equivalence in total islet yield after purification, a significant increase when compared with the control (P = 0.04 and P < 0.01). The islet quality assessments, including an in vivo diabetic nude mice assay and the response of high-mobility group box protein 1 to cytokine stimulation, also showed no significant differences. The proportion of terminal deoxynucleotidyl transferase dUTP nick-end labeling-positive cells showing apoptosis in islets in the PDP groups was significantly lower than in the control group (P < 0.05). Both ET-Kyoto solution and cold storage/purification stock solution are suitable for PDP and consistently resulted in isolation success. Further studies with a larger number of pancreas donors should be done to compare the effects of the PDP solutions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dithizone, Practical Grade
Sigma-Aldrich
Dithizone, ACS reagent, for spectrophotometric det. of Cd, Cu, Hg, Pb, Zn, ≥98.0% (TLC)
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard