Merck
Adverse immunoregulatory effects of 5FU and CPT11 chemotherapy on myeloid-derived suppressor cells and colorectal cancer outcomes.
Cancer research (2014-09-12)
Julia Kanterman, Moshe Sade-Feldman, Moshe Biton, Eliran Ish-Shalom, Audrey Lasry, Aviya Goldshtein, Ayala Hubert, Michal Baniyash
ABSTRAKT
Colorectal cancer is associated with chronic inflammation and immunosuppression mediated by myeloid-derived suppressor cells (MDSC). Although chemotherapy reduces tumor burden at early stages, it tends to have limited effect on a progressive disease, possibly due to adverse effects on the immune system in dictating disease outcome. Here, we show that patients with advanced colorectal cancer display enhanced MDSC levels and reduced CD247 expression and that some conventional colorectal cancer chemotherapy supports the immunosuppressive tumor microenvironment. A FOLFOX combined therapy reduced immunosuppression, whereas a FOLFIRI combined therapy enhanced immunosuppression. Mechanistic studies in a colorectal cancer mouse model revealed that FOLFIRI-like therapy including the drugs CPT11 and 5-fluorouracil (5FU) damaged host immunocompetence in a manner that limits treatment outcomes. CPT11 blocked MDSC apoptosis and myeloid cell differentiation, increasing MDSC immunosuppressive features and mouse mortality. In contrast, 5FU promoted immune recovery and tumor regression. Thus, CPT11 exhibited detrimental immunoregulatory effects that offset 5FU benefits when administered in combination. Our results highlight the importance of developing therapeutic regimens that can target both the immune system and tumor towards improved personalized treatments for colorectal cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar