Merck
Identification of Malassezia species in the facial lesions of Chinese seborrhoeic dermatitis patients based on DNA sequencing.
Mycoses (2014-08-16)
C-h Lian, L-l Shen, Q-y Gao, M Jiang, Z-j Zhao, J-j Zhao
ABSTRAKT
The genus Malassezia is important in the aetiology of facial seborrhoeic dermatitis (FSD), which is the most common clinical type. The purpose of this study was to analyse the distribution of Malassezia species in the facial lesions of Chinese seborrhoeic dermatitis (SD) patients and healthy individuals. Sixty-four isolates of Malassezia were isolated from FSD patients and 60 isolates from healthy individuals. Sequence analysis of the internal transcribed spacer (ITS) region was used to identify the isolates. The most frequently identified Malassezia species associated with FSD was M. furfur (76.56%), followed by M. sympodialis (12.50%) and M. japonica (9.38%). The most frequently isolated species in healthy individuals were M. furfur (61.67%), followed by M. sympodialis (25.00%), M. japonica (6.67%), M. globosa (3.33%), and M. obtusa (3.33%). Overall, our study revealed that while M. furfur is the predominant Malassezia species in Chinese SD patients, there is no significant difference in the distribution of Malassezia species between Chinese SD patients and healthy individuals.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Supelco
3-Methyl-1-butanol, analytical standard
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
3-Methylbutanol, analytical standard
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
3-Methylbutanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%, Molecular Biology
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG