Merck
Next generation sequencing of pancreatic cyst fluid microRNAs from low grade-benign and high grade-invasive lesions.
Cancer letters (2014-10-12)
Jin Wang, Pamela L Paris, Jinyun Chen, Vy Ngo, Hui Yao, Marsha L Frazier, Ann M Killary, Chang-Gong Liu, Han Liang, Christian Mathy, Sandhya Bondada, Kimberly Kirkwood, Subrata Sen
ABSTRAKT
Intraductal papillary mucinous neoplasm (IPMN) is a precursor cystic lesion to pancreatic cancer. With the goal of classifying IPMN cases by risk of progression to pancreatic cancer, we undertook an exploratory next generation sequencing (NGS) based profiling study of miRNAs (miRNome) in the cyst fluids from low grade-benign and high grade-invasive pancreatic cystic lesions. Thirteen miRNAs (miR-138, miR-195, miR-204, miR-216a, miR-217, miR-218, miR-802, miR-155, miR-214, miR-26a, miR-30b, miR-31, and miR-125) were enriched and two miRNAs (miR-451a and miR-4284) were depleted in the cyst fluids derived from invasive carcinomas. Quantitative real-time polymerase chain reaction analysis confirmed that the relative abundance of tumor suppressor miR-216a and miR-217 varied significantly in these cyst fluid samples. Ingenuity Pathway Analysis (IPA) analysis indicated that the genes targeted by the differentially enriched cyst fluid miRNAs are involved in five canonical signaling pathways, including molecular mechanisms of cancer and signaling pathways implicated in colorectal, ovarian and prostate cancers. Our findings make a compelling case for undertaking in-depth analyses of cyst fluid miRNomes for developing informative early detection biomarkers of pancreatic cancer developing from pancreatic cystic lesions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)