Merck
  • Sterically stabilized polymeric nanoparticles with a combinatorial approach for multi drug resistant cancer: in vitro and in vivo investigations.
Sterically stabilized polymeric nanoparticles with a combinatorial approach for multi drug resistant cancer: in vitro and in vivo investigations.
International journal of pharmaceutics (2014-12-03)
Sobiya Zafar, Lalit Mohan Negi, Anita Kamra Verma, Vijay Kumar, Aakriti Tyagi, Pratibha Singh, Zeenat Iqbal, Sushama Talegaonkar
ABSTRAKT
The present work describes the preparation of sterically stabilize polymeric nanoparticles of mitoxantrone dihydrochloride (MTO) along with an efflux transporter (Pgp/BCRP) inhibitor that enhance the circulation time of nanoparticles and simultaneously surmount the problem of multidrug resistance (MDR). Mitoxantrone dihydrochloride being hydrophilic in nature had very low entrapment efficiency (%E.E.), thus in order to further enhance the lipophilicity and the %E.E., it was complexed with sodium deoxycholate (SDC) and this MTO-SDC-complex was used to formulate nanoparticles with/without Pgp/BCRP inhibitor by nanoprecipitation technique and was characterized for various in vitro and in vivo attributes. In vitro cell line studies were conducted on MCF7, A2780(p) and A2780(adr) cells. Furthermore, the targeting potential of hyaluronic acid (HA) coated nanoparticles for CD44 receptors was investigated using the MCF7 cell line. A reduction in the IC50 value observed with the inhibitor loaded nanoparticles in different cell lines indicated the BCRP/Pgp inhibiting ability of the formulated nanoparticles. The reduced macrophage uptake and the increased residence time in blood demonstrated the long circulating behaviour of the nanoparticles. The enhanced cellular uptake of HA coated nanoparticles in MCF7 cells revealed their targeting potential. The HA coated nanoparticles along with efflux transporter inhibitor exhibits a great potential for targeted chemotherapy in CD44 overexpressing MDR breast cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Trioctylamine, 98%
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Kwercetyna, United States Pharmacopeia (USP) Reference Standard
Mitoxantrone hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Bromek cetrymonium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Coumarin 6, ≥99%
Sigma-Aldrich
Hesperetin, ≥95%
Supelco
Hesperetin, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Sigma-Aldrich
Coumarin 6, 98%
Sigma-Aldrich
Methyltrioxorhenium(VII), Re 71.0-76.0 %
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
SAFC
Sodium deoxycholate