Merck
Metabolic profiling of Gynostemma pentaphyllum extract in rat serum, urine and faeces after oral administration.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-26)
Dao-Jin Chen, Hua-Gang Hu, Shao-Fang Xing, Ya-Jun Gao, Si-Fan Xu, Xiang-Lan Piao
ABSTRAKT
Folk drug Gynostemma pentaphyllum (Thunb.) Makino contains many biologically active phytochemicals which have been demonstrated to be effective against chronic diseases. As in vivo anti-tumor experiments of G. pentaphyllum extract (GP) show much stronger antitumor activities than in vitro, it is important and necessary to understand the metabolic study of GP. A sensitive and specific U-HPLC-MS method was utilized for the first time to rapidly identify gypenosides and its possible metabolites in rat serum, urine, and faeces after oral administration. Solid phase extraction was utilized in the sample preparation. Negative Electrospray ionisation (ESI) mass spectrometry was used to discern gypenosides and its possible metabolites in rat samples. As a result, after oral administration, a total of seven metabolites of G. pentaphyllum extract were assigned, two from the rat serum and seven both from the rat urine and faeces. As metabolites of G. pentaphyllum extract, all of them have never been reported before.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%