Merck
Phase I ficlatuzumab monotherapy or with erlotinib for refractory advanced solid tumours and multiple myeloma.
British journal of cancer (2014-06-06)
A Patnaik, G J Weiss, K P Papadopoulos, C C Hofmeister, R Tibes, A Tolcher, R Isaacs, J Jac, M Han, F C Payumo, M M Cotreau, R K Ramanathan
ABSTRAKT
Ficlatuzumab, a humanised hepatocyte growth factor (HGF) IgG1κ inhibitory monoclonal antibody, was evaluated for recommended phase II dose (RP2D), safety, pharmacokinetics (PKs), antidrug antibody (ADA), pharmacodynamics (PDs) and antitumour activity as monotherapy or combined with erlotinib. Patients with solid tumours received ficlatuzumab 2, 5, 10 or 20 mg kg(-1) intravenously every 2 weeks (q2w). Additional patients were treated at the RP2D erlotinib. Forty-one patients enrolled at doses ⩽20 mg kg(-1). Common adverse events (AEs) included peripheral oedema, fatigue and nausea. Three patients experienced grade ⩾3 treatment-related hyperkalaemia/hypokalaemia, diarrhoea or fatigue. Best overall response (44%) was stable disease (SD); median duration was 5.5 months (0.4-18.7 months). One patient has been on therapy with SD for >4 years. Pharmacokinetics of ficlatuzumab showed low clearance (0.17-0.26 ml h(-1) kg(-1)), a half-life of 6.8-9.4 days and dose-proportional exposure. Ficlatuzumab/erlotinib had no impact on the PK of either agent. No ADAs were detected. Ficlatuzumab increased serum HGF levels. Recommended phase II dose is 20 mg kg(-1) q2w for ficlatuzumab monotherapy or with erlotinib. Preliminary antitumour activity and manageable AEs were observed. Pharmacokinetics were dose-proportional and consistent with other IgG therapeutics. Ficlatuzumab was not immunogenic, and serum HGF was a potential PD marker.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%