Merck
Schedule-dependent effects of kappa-selenocarrageenan in combination with epirubicin on hepatocellular carcinoma.
Asian Pacific journal of cancer prevention : APJCP (2014-05-30)
Yu-Bin Ji, Na Ling, Xiao-Jun Zhou, Yun-Xiang Mao, Wen-Lan Li, Ning Chen
ABSTRAKT
Hepatocellular carcinoma (HCC) has a relatively higher incidence in many countries of Asia. Globally, HCC has a high fatality rate and short survival. Epirubicin, a doxorubicin analogue, may be administered alone or in combination with other agents to treat primary liver cancer and metastatic diseases. However, the toxic effects of epirubicin to normal tissues and cells have been one of the major obstacles to successful cancer chemotherapy. Here, we investigated the effects of epirubicin in combination with kappa-selenocarrageenan on mice with H22 implanted tumors and HepG-2 cell proliferation, immune organ index, morphology, cell cycle and related protein expressions in vivo and in vitro with sequential drug exposure. The inhibitory rate of tumor growth in vivo was calculated. Drug sensitivity was measured by MTT assay, and the King's principle was used to evaluate the interaction of drug combination. Morphological changes were observed by fluorescent microscopy. Cell cycle changes were analyzed by flow cytometry. Expression of cyclin A, Cdc25A and Cdk2 were detected by Western blotting. In vivo results demonstrated that the inhibitory rate of EPI combined with KSC was higher than that of KSC or EPI alone, and the Q value indicated an additive effect. In addition, KSC could significantly raise the thymus and spleen indices of mice with H22 implanted tumors. In the drug sensitivity assay in vitro, exposure to KSC and EPI simultaneously was more effective than exposure sequentially in HepG-2 cells, while exposure to KSC prior to EPI was more effective than exposure to EPI prior to KSC. Q values showed an additive effect in the simultaneous group and antagonistic effects in the sequential groups. Morphological analysis showed similar results to the drug sensitivity assay. Cell cycle analysis revealed that exposure to KSC or EPI alone arrested the cells in S phase in HepG-2 cells, exposure to KSC and EPI simultaneously caused accumulation in the S phase, an effect caused by either KSC or EPI. Expression of cyclin A, Cdc25A and Cdk2 protein was down-regulated following exposure to KSC and EPI alone or in combination, exposure to KSC and EPI simultaneously resulting in the lowest values. Taken together, our findings suggest that KSC in combination with EPI might have potential as a new therapeutic regimen against HCC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Selenium, pellets, < 5mm, ≥99.999%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Propidium iodide solution