Merck
Naturally occurring alpha-synuclein autoantibodies in Parkinson's disease: sources of (error) variance in biomarker assays.
PloS one (2014-12-04)
Sebastian Heinzel, Maike Gold, Christian Deuschle, Felix Bernhard, Walter Maetzler, Daniela Berg, Richard Dodel
ABSTRAKT
Alpha-synuclein (α-Syn) plays a pivotal role in the pathophysiology of Parkinson's disease (PD), which can partly be modulated by innate and adaptive immune functions, and vice versa. Here, naturally occurring α-Syn autoantibodies (α-Syn-nAbs) may be effective against α-Syn pathoetiology and may serve as a PD biomarker. However, serum and cerebrospinal fluid α-Syn-nAbs levels still lack consistent evidence as required for a reliable PD biomarker. Serum and cerebrospinal fluid α-Syn-nAbs levels of 66 PD patients and 69 healthy controls were assessed using a validated ELISA assay. Moreover, potential sources of error variance including unspecific ELISA background signals, free serum hemoglobin concentrations, α-Syn plate coating procedures, and differences in α-Syn-nAbs standards, were investigated. PD patients and controls did not differ in serum (p = .49) nor cerebrospinal fluid (p = .29) α-Syn-nAbs levels. Interestingly, free serum hemoglobin concentrations were negatively correlated with α-Syn-nAbs levels in controls (Spearman ρ = -.41, p<.001), but not in PD patients (ρ = .16, p = .21). ELISA α-Syn plate coating procedures impacted inter-assay variability (same day coating: 8-16%; coating on different days: 16-58%). α-Syn-nAbs standards from different purification batches differed regarding optical density measured in ELISAs suggesting differences in α-Syn affinity. While α-Syn-nAbs levels may represent a potential PD biomarker, several methodological issues have to be considered to increase reproducibility of α-Syn-nAbs findings. Further studies using standardized protocols minimizing sources of error variance may be necessary to establish a reliable PD α-Syn-nAbs biomarker.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC