Merck
Hemolytic Streptococcus may exacerbate kidney damage in IgA nephropathy through CCL20 response to the effect of Th17 cells.
PloS one (2014-09-30)
Ting Meng, Xiaozhao Li, Xiang Ao, Yong Zhong, Rong Tang, Weisheng Peng, Jinghua Yang, Mingxiang Zou, Qiaoling Zhou
ABSTRAKT
The exacerbation of IgA nephropathy (IgAN) is related to respiratory tract infection with hemolytic streptococcus (HS), but the mechanism is unknown. In this study we investigated the role of chemokine ligand 20 (CCL20) in response to the effect of T helper 17 (Th17) cells in the pathogenesis of IgAN associated with HS. Thirty mice were randomly divided into five groups: control mice (control), IgAN mice (IgAN), HS-infected IgAN mice (HS-IgAN), CCL20-treated IgAN mice (CCL20-IgAN), and CCL20-treated HS infected IgAN mice (CCL20-HS-IgAN). IgAN mice were induced with lipopolysaccharide, carbon tetrachloride and bovine serum albumin. Then the mice were sensitized with CCL20 antibody and infected with alpha-hemolytic streptococcus (α-HS) isolated from tonsils in sequence. Urine Albumin-Creatinine ratio and sediments were measured. The pathological changes in kidney and lung tissues were observed under microscopy. Th17 cells and regulatory T cells (Tregs) in kidneys were tested by flow cytometry. CCL20, IL-17A, IL-6 and IL-21 in the kidneys were detected by ELISA. The IgAN mice had albuminuria and microscopic hematuria, renal mesangial proliferation, IgA deposition, high electron dense deposition in glomerular mesangial region, decreased frequency of Tregs, increased frequency of Th17 and Th17-Treg ratio. Furthermore, Th17-related cytokines CCL20, IL-17A, IL-6 and IL-21 were all increased in the kidneys of IgAN mice. Compared with IgAN mice, the manifestations in HS-IgAN mice were more severe, but alleviated in CCL20-treated groups. α-HS may exacerbate kidney damage in IgAN through CCL20 response to the effect of Th17 cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material