Merck
PCP4 regulates Purkinje cell excitability and cardiac rhythmicity.
The Journal of clinical investigation (2014-10-09)
Eugene E Kim, Akshay Shekhar, Jia Lu, Xianming Lin, Fang-Yu Liu, Jie Zhang, Mario Delmar, Glenn I Fishman
ABSTRAKT
Cardiac Purkinje cells are important triggers of ventricular arrhythmias associated with heritable and acquired syndromes; however, the mechanisms responsible for this proarrhythmic behavior are incompletely understood. Here, through transcriptional profiling of genetically labeled cardiomyocytes, we identified expression of Purkinje cell protein-4 (Pcp4), a putative regulator of calmodulin and Ca2+/calmodulin-dependent kinase II (CaMKII) signaling, exclusively within the His-Purkinje network. Using Pcp4-null mice and acquired cardiomyopathy models, we determined that reduced expression of PCP4 is associated with CaMKII activation, abnormal electrophysiology, dysregulated intracellular calcium handling, and proarrhythmic behavior in isolated Purkinje cells. Pcp4-null mice also displayed profound autonomic dysregulation and arrhythmic behavior in vivo. Together, these results demonstrate that PCP4 regulates cardiac excitability through both Purkinje cell-autonomous and central mechanisms and identify this modulator of CaMKII signaling as a potential arrhythmia-susceptibility candidate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Przeciwciało anty-HCN4, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Ryanodine Receptor 2 Antibody, serum, Chemicon®
Sigma-Aldrich
DL-Tyrosine, 99%
Tyrosine, European Pharmacopoeia (EP) Reference Standard